5-Fluoro-2-methylbenzaldehyde 97%

Supplier: Ambeed
Warning

A248664-10G A248664-1G A248664-25G
76692-414EA 21 USD
76692-414 76692-416 76692-418
5-Fluoro-2-methylbenzaldehyde 97%
5-Fluoro-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇FO
MDL Number: MFCD02261763
CAS Number: 22062-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR