Boc-Tyr(BZL)-OL 97%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Butyl (1-(4-(benzyloxy)phenyl)-3-hydroxypropan-2-yl)carbamate

A248418-1G A248418-250MG A248418-5G
76692-408EA 38.2 USD
76692-408 76692-410 76692-412
Boc-Tyr(BZL)-OL 97%
Boc-Tyr(BZL)-OL

Precaución: for Research Use Only

Formula: C₂₁H₂₇NO₄
MW: 357.45 g/mol
MDL Number: MFCD00235956
CAS Number: 66605-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR