3-Hydroxy-3-methyl butyric acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxyisovaleric acid, 3-Hydroxy-3-methylbutanoic acid

A246884-100G A246884-500G A246884-5G A246884-25G A246884-1000G
76692-392EA 35 USD
76692-392 76692-394 76692-396 77090-872 77626-304
3-Hydroxy-3-methyl butyric acid 95%
3-Hydroxy-3-methyl butyric acid

Precaución: for Research Use Only

Formula: C₅H₁₀O₃
MW: 118.13 g/mol
MDL Number: MFCD00059081
CAS Number: 625-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR