2-(1-Methylguanidino)ethyl dihydrogen phosphate 98% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: Creatinol phosphate, Aplodan, 2-(1-Methylguanidino)ethyl dihydrogen phosphate

A246801-100G A246801-500G A246801-5G
76692-386EA 26.53 USD
76692-386 76692-388 76692-390
2-(1-Methylguanidino)ethyl dihydrogen phosphate 98% (by HPLC)
2-(1-Methylguanidino)ethyl dihydrogen phosphate

Precaución: for Research Use Only

Formula: C₄H₁₂N₃O₄P
MW: 197.1295 g/mol
MDL Number: MFCD00868297
CAS Number: 6903-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR