Dibenzyl ether 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Benzyl ether
A246600-1000G
A246600-100G
A246600-25G
A246600-500G
76692-378EA
16.98
USD
76692-378
76692-380
76692-382
76692-384
Dibenzyl ether 95%
Dibenzyl ether
Precaución: for Research Use Only
Formula:
C₆H₅CH₂OCH₂C₆H₅ MW: 198.26 g/mol |
MDL Number:
MFCD00004780 CAS Number: 103-50-4 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...