6-tert-Butyl-m-cresol 97%

Supplier: Ambeed
Warning

Synonyms: 2-tert-Butyl-5-methylphenol, 5-Methyl-2-tert-butylphenol

A243395-1000G A243395-100G A243395-500G
76692-336EA 98.67 USD
76692-336 76692-338 76692-340
6-tert-Butyl-m-cresol 97%
6-tert-Butyl-m-cresol

Precaución: for Research Use Only

Formula: C₁₁H₁₆O
MW: 164.25 g/mol
MDL Number: MFCD00002308
CAS Number: 88-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR