Butyl-n-octanoate 99%

Supplier: Ambeed
Warning

A242684-100G A242684-25G A242684-5G
76692-318EA 81 USD
76692-318 76692-320 76692-322
Butyl-n-octanoate 99%
Butyl-n-octanoate

Precaución: for Research Use Only

Formula: C₁₂H₂₄O₂
MDL Number: MFCD00048918
CAS Number: 589-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR