Carbinoxamine Maleate 99+%

Supplier: Ambeed
Danger

Synonyms: Carbinoxamine maleate salt

A242556-100MG A242556-1G A242556-25G A242556-5G
76692-316EA 5 USD
76692-316 77877-811 77877-812 77877-813
Carbinoxamine Maleate 99+%
Carbinoxamine Maleate

Precaución: for Research Use Only

Formula: C₁₆H₁₉ClN₂O·C₄H₄O₄
MW: 406.86 g/mol
MDL Number: MFCD00082461
CAS Number: 3505-38-2
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR