6-Bromo-8-methylquinoline 97%

Supplier: Ambeed
Warning

A241510-250MG A241510-5G A241510-1G
76692-296EA 27.59 USD
76692-296 76692-298 77626-282
6-Bromo-8-methylquinoline 97%
6-Bromo-8-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈BrN
MW: 222.081 g/mol
MDL Number: MFCD11847622
CAS Number: 178396-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR