(R)-4-Phenyloxazolidine-2-thione 98+%

Supplier: Ambeed
Warning

A241359-100MG A241359-1G A241359-250MG
76692-288EA 18.04 USD
76692-288 76692-290 76692-292
(R)-4-Phenyloxazolidine-2-thione 98+%
(R)-4-Phenyloxazolidine-2-thione

Precaución: for Research Use Only

Formula: C₉H₉NOS
MW: 179.23 g/mol
MDL Number: MFCD06658220
CAS Number: 171877-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR