3-Fluoro-4-methylbenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A239558-1G
A239558-25G
A239558-5G
A239558-100G
76692-250EA
8
USD
76692-250
76692-252
76692-254
77877-798
3-Fluoro-4-methylbenzaldehyde 97%
3-Fluoro-4-methylbenzaldehyde
Precaución: for Research Use Only
Formula:
C₈H₇FO |
MDL Number:
MFCD01631532 CAS Number: 177756-62-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...