Canertinib dihydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: N-[4-[(3-Chloro-4-fluorophenyl)amino]-7-[3-(4-morpholinyl)propoxy]-6-quinazolinyl]-2-propenamide dihydrochloride, CI-1033, PD-183805

A239262-10MG A239262-1MG A239262-50MG
76692-242EA 51 USD
76692-242 76692-244 76692-246 76692-248
Canertinib dihydrochloride 98%
Canertinib dihydrochloride

Precaución: for Research Use Only

Formula: C₂₄H₂₅ClFN₅O₃·2HCl
MW: 558.86 g/mol
MDL Number: MFCD09954112
CAS Number: 289499-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR