rel-4-Amino-5-chloro-N-((3R,4S)-1-(3-(4-fluorophenoxy)propyl)-3-methoxypiperidin-4-yl)-2-methoxybenzamide 98%

Supplier: Ambeed
Warning

Synonyms: Cisapride

A238567-10MG A238567-250MG A238567-25G A238567-50MG A238567-5G A238567-1G
76692-232EA 5 USD
76692-232 76692-234 76692-236 76692-238 76692-240 77600-918
rel-4-Amino-5-chloro-N-((3R,4S)-1-(3-(4-fluorophenoxy)propyl)-3-methoxypiperidin-4-yl)-2-methoxybenzamide 98%
rel-4-Amino-5-chloro-N-((3R,4S)-1-(3-(4-fluorophenoxy)propyl)-3-methoxypiperidin-4-yl)-2-methoxybenzamide

Precaución: for Research Use Only

Formula: C₂₃H₂₉ClFN₃O₄
MW: 465.94 g/mol
MDL Number: MFCD00869327
CAS Number: 81098-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR