Benzyl-(S)-(-)-lactate 98%

Supplier: Ambeed
Warning

Synonyms: Benzyl L-lactate, (S)-(-)-Lactic Acid Benzyl Ester, Benzyl (S)-Lactate

A238307-100G A238307-10G A238307-1G A238307-25G A238307-5G
76692-222EA 535.76 USD
76692-222 76692-224 76692-226 76692-228 76692-230
Benzyl-(S)-(-)-lactate 98%
Benzyl-(S)-(-)-lactate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.20 g/mol
MDL Number: MFCD00209546
CAS Number: 56777-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR