Sodium 8-hydroxy-7-((6-sulfonatonaphthalen-2-yl)diazenyl)quinoline-5-sulfonate 95%

Supplier: Ambeed
Warning

Synonyms: NSC-87877 disodium

A236181-10MG A236181-1MG A236181-25MG A236181-50MG A236181-5MG
76692-170EA 45.62 USD
76692-170 76692-172 76692-174 76692-176 76692-178
Sodium 8-hydroxy-7-((6-sulfonatonaphthalen-2-yl)diazenyl)quinoline-5-sulfonate 95%
Sodium 8-hydroxy-7-((6-sulfonatonaphthalen-2-yl)diazenyl)quinoline-5-sulfonate

Precaución: for Research Use Only

Formula: C₁₉H₁₁N₃Na₂O₇S₂
MW: 503.42 g/mol
MDL Number: MFCD00041863
CAS Number: 56932-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR