Methyl 2-(tert-Butoxycarbonylamino)-3-nitrobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Boc-amino)-3-nitrobenzoic acid methyl ester, 2-(tert-Butoxycarbonylamino)-3-nitrobenzoic acid methyl ester, Methyl 2-(Boc-amino)-3-nitrobenzoate, Methyl 2-((tert-butoxycarbonyl)amino)-3-nitrobenzoate

A235587-100G A235587-1G A235587-25G A235587-5G
76692-148EA 721 USD
76692-148 76692-150 76692-152 76692-154
Methyl 2-(tert-Butoxycarbonylamino)-3-nitrobenzoate 98%
Methyl 2-(tert-Butoxycarbonylamino)-3-nitrobenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂O₆
MDL Number: MFCD06797690
CAS Number: 57113-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR