4-Methoxyphenylacetone 98%

Supplier: Ambeed
Warning

A234955-1000G A234955-100G A234955-10G A234955-25G A234955-500G A234955-5G
76692-128EA 234 USD
76692-128 76692-130 76692-132 76692-134 76692-136 76692-138
4-Methoxyphenylacetone 98%
4-Methoxyphenylacetone

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MDL Number: MFCD00008773
CAS Number: 122-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR