Methyl 4-[(chlorosulfonyl)methyl]benzoate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Methyl 4-((chlorosulphonyl)methyl)benzoate, [4-(Methoxycarbonyl)phenyl]methanesulphonyl chloride, Methyl 4-(chlorosulphonylmethyl)benzoate
A234504-100MG
A234504-250MG
76692-120EA
51
USD
76692-120
77877-787
Methyl 4-[(chlorosulfonyl)methyl]benzoate 98%
Methyl 4-[(chlorosulfonyl)methyl]benzoate
Precaución: for Research Use Only
Formula:
C₉H₉ClO₄S MW: 248.683 g/mol |
MDL Number:
MFCD07778383 CAS Number: 130047-14-2 UN: 1759 ADR: 8 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...