Cinnamoyl chloride 97% predominantly trans

Supplier: Ambeed
Danger

A234326-25G A234326-100G
76692-118EA 7 USD
76692-118 77600-886
Cinnamoyl chloride 97% predominantly trans
Cinnamoyl chloride

Precaución: for Research Use Only

Formula: C₉H₇ClO
MDL Number: MFCD00000732
CAS Number: 102-92-1
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR