Cyclohexyl phenyl ketone 98%

Supplier: Ambeed
Warning

A233691-100G A233691-25G A233691-5G A233691-500G
76692-096EA 109 USD
76692-096 76692-098 76692-100 77626-236
Cyclohexyl phenyl ketone 98%
Cyclohexyl phenyl ketone

Precaución: for Research Use Only

Formula: C₁₃H₁₆O
MDL Number: MFCD00001467
CAS Number: 712-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR