Cyclopropyl-4-fluorophenyl ketone 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
Cyclopropyl(4-fluorophenyl)methanone
A232628-10G
A232628-1G
A232628-250MG
A232628-25G
A232628-5G
76692-088EA
76.39
USD
76692-088
76692-090
76692-092
76692-094
77626-228
Cyclopropyl-4-fluorophenyl ketone 97%
Cyclopropyl-4-fluorophenyl ketone
Precaución: for Research Use Only
Formula:
C₁₀H₉FO MW: 164.18 g/mol |
MDL Number:
MFCD00013728 CAS Number: 772-31-6 UN: 1993 ADR: 3,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...