cis-1,2,3,6-Tetrahydrophthalimide 98%

Supplier: Ambeed
Warning

Synonyms: cis-4-Cyclohexene-1,2-dicarboximide, cis-3a,4,7,7a-Tetrahydro-1H-isoindole-1,3(2H)-dione

A232490-1000G A232490-100G A232490-10G A232490-25G A232490-500G
76692-078EA 115.64 USD
76692-078 76692-080 76692-082 76692-084 76692-086
cis-1,2,3,6-Tetrahydrophthalimide 98%
cis-1,2,3,6-Tetrahydrophthalimide

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.17 g/mol
MDL Number: MFCD00005880
CAS Number: 1469-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR