Boc-Phe-ONp 97%

Supplier: Ambeed
Warning

Synonyms: N-Boc-L-phenylalanine 4-nitrophenyl ester

A231194-10G A231194-25G A231194-100G A231194-5G
76692-068EA 22.28 USD
76692-068 77600-850 77626-218 77626-220
Boc-Phe-ONp 97%
Boc-Phe-ONp

Precaución: for Research Use Only

Formula: C₂₀H₂₂N₂O₆
MW: 386.404 g/mol
MDL Number: MFCD00042811
CAS Number: 7535-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR