2,2,2-Trifluoro-3'-(trifluoromethyl)acetophenone 95%

Supplier: Ambeed
Danger

Synonyms: 2,2,2-Trifluoro-1-(3-(trifluoromethyl)phenyl)ethanone

A230840-1G A230840-5G
76692-064EA 41.38 USD
76692-064 76692-066
2,2,2-Trifluoro-3'-(trifluoromethyl)acetophenone 95%
2,2,2-Trifluoro-3'-(trifluoromethyl)acetophenone

Precaución: for Research Use Only

Formula: C₉H₄F₆O
MW: 242.12 g/mol
MDL Number: MFCD00061275
CAS Number: 721-37-9
UN: 1224
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR