Benzyl laurate 98%

Supplier: Ambeed
Warning

Synonyms: Benzyl dodecanoate, Lauric Acid Benzyl Ester, Benzyl Laurate

A230354-100G A230354-5G
76692-054EA 185 USD
76692-054 76692-056
Benzyl laurate 98%
Benzyl laurate

Precaución: for Research Use Only

Formula: C₁₉H₃₀O₂
MW: 290.45 g/mol
MDL Number: MFCD00026558
CAS Number: 140-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR