Atractyloside potassium salt 95%

Supplier: Ambeed
Danger

Synonyms: Potassium (2R,4aR,6aR,7S,9R,11aS,11bR)-7-hydroxy-2-(((2R,3R,4R,5R,6R)-6-(hydroxymethyl)-3-((3-methylbutanoyl)oxy)-5-(sulphonatooxy)-4-(sulphooxy)tetrahydro-2H-pyran-2-yl)oxy)-11b-methyl-8-methylenetetradecahydro-6a,9-methanocyclohepta[a]naphthalene-4-carbox, (2β)-15α-hydroxy-2-[[2-O-(3-methyl-1-oxobutyl)-3,4-di-O-sulpho-β-D-glucopyranosyl]oxy]-19-norkaur-16-en-18-oic acid, dipotassium salt

A230303-10MG A230303-1MG A230303-5MG
76692-048EA 158.08 USD
76692-048 76692-050 76692-052
Atractyloside potassium salt 95%
Atractyloside potassium salt

Precaución: for Research Use Only

Formula: C₃₀H₄₄K₂O₁₆S₂
MW: 802.99 g/mol
MDL Number: MFCD00078810
CAS Number: 102130-43-8
UN: 2811
ADR: 6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR