Benzyl ethyl piperidine-1,3-dicarboxylate 96%

Supplier: Ambeed
Warning

Synonyms: 1-Benzyl 3-ethyl piperidine-1,3-dicarboxylate

A229668-10G A229668-1G A229668-25G A229668-5G A229668-100G
76692-040EA 13.8 USD
76692-040 76692-042 76692-044 76692-046 77593-486
Benzyl ethyl piperidine-1,3-dicarboxylate 96%
Benzyl ethyl piperidine-1,3-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₆H₂₁NO₄
MW: 291.342 g/mol
MDL Number: MFCD06657752
CAS Number: 310454-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR