BAPTA-AM 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tetrakis(acetoxymethyl)-1,2-bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetate, Tetrakis(acetoxymethyl) 1,2-Bis(2-aminophenoxy)ethane-N,N,N,N-tetraacetate, 1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic Acid Tetrakis(acetoxymethyl) Ester, BAPTA-AM ester
A229213-10MG
A229213-1MG
A229213-5MG
A229213-100MG
76692-032EA
29.71
USD
76692-032
76692-034
76692-036
77626-208
BAPTA-AM 95%
BAPTA-AM
Precaución: for Research Use Only
Formula:
C₃₄H₄₀N₂O₁₈ MW: 764.69 g/mol |
MDL Number:
MFCD00036696 CAS Number: 126150-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...