Bis(2-benzamidophenyl)disulfide 98%

Supplier: Ambeed
Warning

Synonyms: 2,2'-Dithiodibenzanilide, 2,2'-Dibenzamidodiphenyl Disulphide, Bis(2-benzamidophenyl) Disulphide

A229128-100G A229128-25G
76692-030EA 26 USD
76692-030 77877-753
Bis(2-benzamidophenyl)disulfide 98%
Bis(2-benzamidophenyl)disulfide

Precaución: for Research Use Only

Formula: C₂₆H₂₀N₂O₂S₂
MDL Number: MFCD00043806
CAS Number: 135-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR