(6-Chloro-pyridin-3-yl)carbamic acid tert-butyl ester 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (6-chloropyridin-3-yl)carbamate

A228692-100G A228692-10G A228692-1G A228692-250MG A228692-25G A228692-5G
76692-018EA 425.43 USD
76692-018 76692-020 76692-022 76692-024 76692-026 76692-028
(6-Chloro-pyridin-3-yl)carbamic acid tert-butyl ester 95%
(6-Chloro-pyridin-3-yl)carbamic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₃ClN₂O₂
MW: 228.68 g/mol
MDL Number: MFCD08062421
CAS Number: 171178-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR