2-Methyl-5-nitroanisole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methoxy-2-methyl-5-nitrobenzene, Methyl 5-nitro-o-tolyl ether, 2-Methoxy-4-nitrotoluene

A227948-100G A227948-10G A227948-25G A227948-5G
76692-010EA 33.95 USD
76692-010 76692-012 76692-014 76692-016
2-Methyl-5-nitroanisole 98%
2-Methyl-5-nitroanisole

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.16 g/mol
MDL Number: MFCD00043912
CAS Number: 13120-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR