Allyl methyl carbonate 98% stabilized
Supplier: Ambeed
|
Danger
|
Synonyms:
Carbonic acid Allyl Methyl Ester, Allyl Methyl Carbonate
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
A227683-25G
A227683-5G
76692-006EA
68.96
USD
76692-006
76692-008
Allyl methyl carbonate 98% stabilized
Allyl methyl carbonate
Precaución: for Research Use Only
Formula:
C₅H₈O₃ MW: 116.1152 g/mol |
MDL Number:
MFCD00134545 CAS Number: 35466-83-2 UN: 3272 ADR: 3,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...