(S)-4-Phenylthiazolidine-2-thione 98%

Supplier: Ambeed
Warning

A227326-100MG A227326-1G A227326-250MG
76691-994EA 21.22 USD
76691-994 76691-996 76691-998
(S)-4-Phenylthiazolidine-2-thione 98%
(S)-4-Phenylthiazolidine-2-thione

Precaución: for Research Use Only

Formula: C₉H₉NS₂
MW: 195.30 g/mol
MDL Number: MFCD06658214
CAS Number: 185137-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR