3,3,5-Trimethylcyclohexyl mandelate (mixture of isomers) 94+%

Supplier: Ambeed
Warning

Synonyms: Cyclandelate, 3,3,5-Trimethylcyclohexyl 2-hydroxy-2-phenylacetate, Mandelic Acid 3,3,5-Trimethylcyclohexyl Ester

A227272-100MG A227272-1G A227272-50MG A227272-5G
76691-986EA 6 USD
76691-986 76691-988 76691-990 76691-992
3,3,5-Trimethylcyclohexyl mandelate (mixture of isomers) 94+%
3,3,5-Trimethylcyclohexyl mandelate (mixture of isomers)

Precaución: for Research Use Only

Formula: C₁₇H₂₄O₃
MDL Number: MFCD00056623
CAS Number: 456-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR