8-Chloro[1,2,4]triazolo[4,3-a]pyrazine 97%

Supplier: Ambeed
Warning

A227030-10G A227030-250MG A227030-1G A227030-5G
76691-974EA 110 USD
76691-974 76691-976 77626-170 77877-731
8-Chloro[1,2,4]triazolo[4,3-a]pyrazine 97%
8-Chloro[1,2,4]triazolo[4,3-a]pyrazine

Precaución: for Research Use Only

Formula: C₅H₃ClN₄
MDL Number: MFCD08272086
CAS Number: 68774-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR