N-(5-Chloro-4-((4-chlorophenyl)(cyano)methyl)-2-methylphenyl)-2-hydroxy-3,5-diiodobenzamide, sodium salt 98%

Supplier: Ambeed
Warning

Synonyms: Closantel sodium

A226988-100G A226988-1G A226988-250MG A226988-25G A226988-500G A226988-5G A226988-100MG
76691-962EA 84 USD
76691-962 76691-964 76691-966 76691-968 76691-970 76691-972 77879-376
N-(5-Chloro-4-((4-chlorophenyl)(cyano)methyl)-2-methylphenyl)-2-hydroxy-3,5-diiodobenzamide, sodium salt 98%
N-(5-Chloro-4-((4-chlorophenyl)(cyano)methyl)-2-methylphenyl)-2-hydroxy-3,5-diiodobenzamide, sodium salt

Precaución: for Research Use Only

Formula: C₂₂H₁₄Cl₂I₂N₂NaO₂
MDL Number: MFCD03427328
CAS Number: 61438-64-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR