Bis(vinylsulfonyl)methane 98%

Supplier: Ambeed
Danger

A226770-1G A226770-25G A226770-5G
76691-952EA 13 USD
76691-952 76691-954 77626-168
Bis(vinylsulfonyl)methane 98%
Bis(vinylsulfonyl)methane

Precaución: for Research Use Only

Formula: C₅H₈O₄S₂
MDL Number: MFCD03265421
CAS Number: 3278-22-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR