(2S)-(+)-2,3-Epoxypropyl toluene-4-sulfonate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
(2S)-(+)-Glycidyl tosylate, (s)-(+)-oxirane-2-methanol p-toluenesulphonate, (2S)-(+)-Glycidyl p-toluenesulphonate
A226221-1G
A226221-5G
A226221-10G
A226221-25G
76691-934EA
5.31
USD
76691-934
76691-936
77626-160
77626-162
(2S)-(+)-2,3-Epoxypropyl toluene-4-sulfonate 98%
(2S)-(+)-2,3-Epoxypropyl toluene-4-sulphonate
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O₄S MW: 228.26 g/mol |
MDL Number:
MFCD00064489 CAS Number: 70987-78-9 UN: 3077 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...