Calcium bis(2,5-dihydroxybenzenesulfonate) 99%

Supplier: Ambeed
Warning

Synonyms: Calcium 2,5-dihydroxybenzenesulphonate, Calcium dobesilate

A225604-10G A225604-25G A225604-500G
76691-928EA 5.31 USD
76691-928 76691-930 76691-932
Calcium bis(2,5-dihydroxybenzenesulfonate) 99%
Calcium bis(2,5-dihydroxybenzenesulfonate)

Precaución: for Research Use Only

Formula: C₁₂H₁₀CaO₁₀S₂
MW: 418.42 g/mol
MDL Number: MFCD00867249
CAS Number: 20123-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR