Butyl-4-hydroxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Butyl 4-hydroxybenzoate, 4-Hydroxybenzoic acid butyl ester, Butyl parahydroxybenzoate, Butylparaben

A225198-1000G A225198-100G A225198-25G A225198-500G A225198-5G
76691-918EA 65.78 USD
76691-918 76691-920 76691-922 76691-924 76691-926
Butyl-4-hydroxybenzoate 98%
Butyl-4-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
MDL Number: MFCD00016478
CAS Number: 94-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR