Benzoxazole 98%

Supplier: Ambeed
Warning

Synonyms: 1,3-Benzoxazole

A224260-100G A224260-10G A224260-25G A224260-500G A224260-5G
76691-892EA 53.05 USD
76691-892 76691-894 76691-896 76691-898 76691-900
Benzoxazole 98%
Benzoxazole

Precaución: for Research Use Only

Formula: C₇H₅NO
MW: 119.12 g/mol
MDL Number: MFCD00005765
CAS Number: 273-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR