Benzo-18-crown-6 (benzo-1,4,7,10,13,16-hexoxacyclooctadecane) 96%

Supplier: Ambeed
Warning

A223957-1G A223957-5G A223957-25G
76691-886EA 40 USD
76691-886 76691-888 77626-134
Benzo-18-crown-6 (benzo-1,4,7,10,13,16-hexoxacyclooctadecane) 96%
Benzo-18-crown-6 (benzo-1,4,7,10,13,16-hexoxacyclooctadecane)

Precaución: for Research Use Only

Formula: C₁₆H₂₄O₆
MDL Number: MFCD00062741
CAS Number: 14098-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR