(R)-Methyl-2-acetamidopropanoate 95%

Supplier: Ambeed
Warning

Synonyms: (R)-Methyl 2-acetamidopropanoate, Ac-D-Ala-OMe

A223290-10G A223290-250MG A223290-5G A223290-1G
76691-852EA 102.91 USD
76691-852 76691-854 77600-786 77626-124
(R)-Methyl-2-acetamidopropanoate 95%
(R)-Methyl-2-acetamidopropanoate

Precaución: for Research Use Only

Formula: C₆H₁₁NO₃
MW: 145.158 g/mol
MDL Number: MFCD00080822
CAS Number: 19914-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR