2'-Deoxy-5-methylcytidine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidin-2(1H)-one, 5-mdC, 5-Methyl-2'-deoxycytidine, 1-(2-Deoxy-beta-D-ribofuranosyl)-5-methylcytosine

A223184-10G A223184-250MG A223184-1G A223184-5G
76691-842EA 304.48 USD
76691-842 76691-844 77600-784 77626-120
2'-Deoxy-5-methylcytidine 98%
2'-Deoxy-5-methylcytidine

Precaución: for Research Use Only

Formula: C₁₀H₁₅N₃O₄
MW: 241.25 g/mol
MDL Number: MFCD00006549
CAS Number: 838-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR