Cinnamyl isobutyrate 98%

Supplier: Ambeed
Warning

A222884-100G A222884-25G
76691-838EA 35 USD
76691-838 76691-840
Cinnamyl isobutyrate 98%
Cinnamyl isobutyrate

Precaución: for Research Use Only

Formula: C₁₃H₁₆O₂
MW: 204.26 g/mol
MDL Number: MFCD00048316
CAS Number: 103-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR