6-Phenylhexan-2-one 95%

Supplier: Ambeed
Warning

A221076-100MG A221076-1G A221076-250MG
76691-820EA 72.15 USD
76691-820 76691-822 76691-824
6-Phenylhexan-2-one 95%
6-Phenylhexan-2-one

Precaución: for Research Use Only

Formula: C₁₂H₁₆O
MW: 176.25 g/mol
MDL Number: MFCD09032287
CAS Number: 14171-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR