Benzo[c]phenanthrene 97%

Supplier: Ambeed
Warning

A220948-100MG A220948-1G A220948-250MG A220948-5G
76691-814EA 8 USD
76691-814 76691-816 76691-818 77877-687
Benzo[c]phenanthrene 97%
Benzo[c]phenanthrene

Precaución: for Research Use Only

Formula: C₁₈H₁₂
MDL Number: MFCD00019126
CAS Number: 195-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR