2-(Dimethylamino)-2-phenylbutan-1-ol 95%

Supplier: Ambeed
Warning

A220538-1G A220538-250MG A220538-25G A220538-5G
76691-806EA 9 USD
76691-806 76691-808 76691-810 76691-812
2-(Dimethylamino)-2-phenylbutan-1-ol 95%
2-(Dimethylamino)-2-phenylbutan-1-ol

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO
MDL Number: MFCD00869525
CAS Number: 39068-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR