Ethyl-7-bromo-1H-indole-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 7-bromo-1H-indole-2-carboxylate

A220323-100G A220323-100MG A220323-10G A220323-1G A220323-250MG A220323-25G A220323-5G
76691-792EA 887.98 USD
76691-792 76691-794 76691-796 76691-798 76691-800 76691-802 76691-804
Ethyl-7-bromo-1H-indole-2-carboxylate 98%
Ethyl-7-bromo-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrNO₂
MW: 268.1066 g/mol
MDL Number: MFCD04972046
CAS Number: 16732-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR