Boc-D-phenylalanine methyl ester 98%

Supplier: Ambeed
Warning

Synonyms: (R)-Methyl 2-((tert-butoxycarbonyl)amino)-3-phenylpropanoate, Boc-D-Phe-OMe, N-Boc-D-phenylalanine methyl ester

A220222-1G A220222-250MG A220222-5G A220222-25G
76691-784EA 13 USD
76691-784 76691-786 76691-788 77600-772
Boc-D-phenylalanine methyl ester 98%
Boc-D-phenylalanine methyl ester

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₄
MW: 279.336 g/mol
MDL Number: MFCD00672509
CAS Number: 77119-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR